Mercury, Oil based standard solution, Specpure®, Hg 1000µg/g, Thermo Scientific

Supplier: Thermo Scientific Chemicals

043756-18
AA43756-18EA 99.9 USD
AA43756-18
Mercury, Oil based standard solution, Specpure®, Hg 1000µg/g, Thermo Scientific
Standards Cationic Standards Mercury Standards

Physical Form: Liquid
MDL No.: MFCD00011035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR