Silicon dioxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
043736-14
043736-22
AA43736-14EA
90.9
USD
AA43736-14
AA43736-22
Silicon dioxide ≥99.99% (metals basis)
Silicium dioxide
For vacuum deposition
Purity: 99.99%
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: 1-3mm Fused Lump
Density: 2.2-2.6
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232
For vacuum deposition
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.2…2.6 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,8493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...