Iodomethane 99+% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl iodide

Stabilization: Stabilised with copper

043681-18 043681-30 043681-A1
AA43681-18EA 42.66 USD
AA43681-18 AA43681-30 AA43681-A1
Iodomethane 99+% stabilised
Iodomethane

Fieser: 1,682 2,274 3,202 4,341 5,447 6,384 8,342 9,308 19,215


Purity: 99+% CAS No.: 74-88-4 Molecular Formula: CH3I Formula Weight: 141.94 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 2.28 Refractive Index: 1.5300 Sensitivity: Light Sensitive Appearance: Colorless to pale yellow Odor: Pungent Melting Point: -67° Boiling Point: 41-43° Beilstein Reference: 969135 Merck Reference: 14,6087 UN No.: 2644 MDL No.: MFCD00001073

Formula: CH₃I
Boiling Pt: 41…43 °C
Melting Pt: –67 °C
Density: 2.28
Storage Temperature: Refrigerator
MDL Number: MFCD00001073
CAS Number: 74-88-4
EINECS: 200-819-5
UN: 2644
ADR: 6.1,I
Merck Index: 14,6087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR