Polyethylene oxide, M.W. 1,000,000

Supplier: Thermo Scientific Chemicals

Synonyms: PEO

043678-14 043678-22 043678-36
AA43678-14EA 45.39 USD
AA43678-14 AA43678-22 AA43678-36
Polyethylene oxide, M.W. 1,000,000
Polyethylene oxide

Inhibited with 200ppm BHT


CAS No.: 25322-68-3 Molecular Formula: (-CH2CH2O-)n Physical Form: Powder Odor: Odorless pH: 5-7 MDL No.: MFCD00147971

Formula: (-CH₂CH₂O-)n
Storage Temperature: Ambient
MDL Number: MFCD00147970
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR