Graphite, rods, pyrolytic coated, Length 152 mm (6 in), Diameter 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

043670-KG 043670-KJ
AA43670-KGEA 123 USD
AA43670-KG AA43670-KJ
Graphite, rods, pyrolytic coated, Length 152 mm (6 in), Diameter 2 mm (0.08 in)
Graphite

Mount marks may be visible after coating


Physical Form: ≈0.8g/pc MDL No.: MFCD00144065

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR