Scandium ≥99.6% (metals basis excluding Ta), rods, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

043499-DH 043499-HF
AA43499-DHEA 1580 USD
AA43499-DH AA43499-HF
Scandium ≥99.6% (metals basis excluding Ta), rods, Diameter 12.7 mm (0.5 in)
Scandium

MDL: MFCD00016323


Purity: 99.6% CAS No.: 7440-20-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.53g/mm, Cast rod, diameter is approximate Appearance: Grey Odor: Odorless MDL No.: MFCD00016323

Formula: Sc
Boiling Pt: 2730 °C (1013 hPa)
Melting Pt: 1541 °C
Density: 2.989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR