Glycine ≥98.5% ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
043497-36
043497-A5
043497-22
AA43497-36EA
144.41
USD
AA43497-36
AA43497-A5
AA43497-22
Glycine ≥98.5% ACS
Glycine
MDL: MFCD00008131
Beilstein Registry No.: 635782
Fieser: 1,412
Purity: 98.5+%
CAS No.: 56-40-6
Molecular Formula: NH2CH2CO2H
Formula Weight: 75.07
Physical Form: Crystalline
Density: 1.595
Appearance: White to off-white
Odor: Odorless
pH: 5,97
Melting Point: ca 245° dec.
Beilstein Reference: 635782
Merck Reference: 14,4491
MDL No.: MFCD00008131
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: ∼ 245 °C (dec.) Density: 1.595 Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 14,4491 |
|
Specification Test Results
Residue after ignition | 0.1 |
Heavy metals (as Pb) | 0.2% (as Pb) |
Cl | 0.005 AU |
SO3 | 0.005% |
NH3 | 0.005% |
Substances darkened by sulfuric acid | Pass test |
Hydrolyzable substances | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...