Tin ≥99.995% (metals basis), slug, Puratronic®, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

043416-22 043416-14
AA43416-22EA 218 USD
AA43416-22 AA43416-14
Tin ≥99.995% (metals basis), slug, Puratronic®, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Tin

MDL: MFCD00133862


Purity: 99.995% CAS No.: 7440-31-5 Formula Weight: 118.710 Physical Form: ≈1.5g/slug Density: 7.310G/CC Odor: Odorless Boiling Point: 2270 deg C MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.310G/CC
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR