Tin ≥99.995% (metals basis), slug, Puratronic®, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
043416-22
043416-14
AA43416-22EA
218
USD
AA43416-22
AA43416-14
Tin ≥99.995% (metals basis), slug, Puratronic®, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Tin
MDL: MFCD00133862
Purity: 99.995%
CAS No.: 7440-31-5
Formula Weight: 118.710
Physical Form: ≈1.5g/slug
Density: 7.310G/CC
Odor: Odorless
Boiling Point: 2270 deg C
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C Melting Pt: 231.9 °C Density: 7.310G/CC Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...