Indium ≥99.998% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

043414-14 043414-22
AA43414-14EA 243 USD
AA43414-14 AA43414-22
Indium ≥99.998% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Indium

MDL: MFCD00134048


Purity: 99.998% CAS No.: 7440-74-6 Formula Weight: 114.818 Physical Form: ≈1.5g/slug Density: 7.310G/CC Odor: Odorless Boiling Point: 2072 deg C MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.310G/CC
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR