Methyl levulinate 99+%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
4-Ketovaleric acid methyl ester, 4-Oxopentanoic acid methyl ester, Methyl 4-oxopentanoate, 4-Oxovaleric acid methyl ester, Methyl 4-oxovalerate, Levulinic acid methyl ester
043397-14
043397-22
AA43397-36EA
737.72
USD
AA43397-36
AA43397-14
AA43397-22
Methyl levulinate 99+%
Methyl levulinate
MDL: MFCD00017499
Purity: 99+%
CAS No.: 624-45-3
Molecular Formula: CH3COCH2CH2CO2CH3
Formula Weight: 130.14
Flash Point: 72°(161.6°F)
Physical Form: Liquid
Density: 1.05
Odor: Pleasant
Boiling Point: 191°
MDL No.: MFCD00017499
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol Boiling Pt: 191 °C Melting Pt: -24 Density: 1.05 Flash Pt: 72 °C (161.6 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017499 CAS Number: 624-45-3 EINECS: 210-846-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...