Arsenic (III) sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Arsenic trisulphide, Arsenic trisulfide, Disrsenic trisulfide, Disrsenic trisulphide

043340-06 043340-14
AA43340-06EA 64.8 USD
AA43340-06 AA43340-14
Arsenic (III) sulfide ≥99.9% (metals basis)
Arsenic (III) sulphide

For vacuum deposition


Purity: 99.9% CAS No.: 1303-33-9 Molecular Formula: As2S3 Formula Weight: 246.03 Physical Form: 1-6mm Fused Pieces Density: 3.43 Melting Point: 300° Boiling Point: 707° Merck Reference: 14,806 UN No.: 1557 MDL No.: MFCD00003435


For vacuum deposition

Formula: As₂S₃
Boiling Pt: 707 °C
Melting Pt: 300 °C
Density: 3.43
Storage Temperature: Ambient
MDL Number: MFCD00003435
CAS Number: 1303-33-9
EINECS: 215-117-4
UN: 1557
ADR: 6.1,III
Merck Index: 14,806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR