Silicon carbide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
043332-36
043332-A3
AA43332-36EA
134
USD
AA43332-36
AA43332-A3
Silicon carbide ≥99% (metals basis)
Silicon carbide
MDL: MFCD00049531
Purity: 99%
CAS No.: 409-21-2
Molecular Formula: CSi
Formula Weight: 40.10
Physical Form: -325 Mesh Powder, Black
Density: 3.16
Appearance: Black
Odor: Odorless
Melting Point: 2700°
Merck Reference: 14,8492
MDL No.: MFCD00049531
Formula:
SiC Melting Pt: 2700 °C Density: 3.16 Storage Temperature: Ambient |
MDL Number:
MFCD00049531 CAS Number: 409-21-2 EINECS: 206-991-8 Merck Index: 14,8492 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...