n-Hexadecane ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexadecane, Cetane
043283-LV
043283-LW
AA43283-LVEA
187.7
USD
AA43283-LV
AA43283-LW
n-Hexadecane ≥95%
n-Hexadecane
Standard for rating diesel-fuel ignition qualities
Purity: 95%
CAS No.: 544-76-3
Molecular Formula: CH3(CH2)14CH3
Formula Weight: 226.45
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4345
Appearance: Colorless
Melting Point: 18°
Boiling Point: 287°
Solubility: Soluble in ethanol
Beilstein Reference: 1736592
MDL No.: MFCD00008998
Standard for rating diesel-fuel ignition qualities
Formula:
H₃C(CH₂)₁₄CH₃ Boiling Pt: 287 °C Melting Pt: 18 °C Density: 0.773 Flash Pt: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008998 CAS Number: 544-76-3 EINECS: 208-878-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...