Indium ≥99.99% (metals basis), wire, Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

043265-BU 043265-G5 043265-G1
AA43265-BUEA 84.2 USD
AA43265-BU AA43265-G5 AA43265-G1
Indium ≥99.99% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Indium

MDL: MFCD00134048


Purity: 99.99% CAS No.: 7440-74-6 Physical Form: ≈22.95g/m Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR