Indium ≥99.99% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
043265-BU
043265-G5
043265-G1
AA43265-BUEA
84.2
USD
AA43265-BU
AA43265-G5
AA43265-G1
Indium ≥99.99% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Indium
MDL: MFCD00134048
Purity: 99.99%
CAS No.: 7440-74-6
Physical Form: ≈22.95g/m
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...