Acetyl chloride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetyl chloride 1M in Dichloromethane
043262-AD
043262-AK
043262-K2
AA43262-ADEA
52.18
USD
AA43262-AD
AA43262-AK
AA43262-K2
Acetyl chloride 99+%
Acetyl chloride
Purity: 99+%
CAS No.: 75-36-5
Molecular Formula: C2H3ClO
Formula Weight: 78.50
Flash Point: 4°(39°F)
Physical Form: Liquid
Density: 1.104
Refractive Index: 1.3890
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Pungent
Melting Point: -112°
Boiling Point: 50-52°
Beilstein Reference: 605303
Merck Reference: 14,85
UN No.: 1717
MDL No.: MFCD00000719
Formula:
C₂H₃ClO Boiling Pt: 50…52 °C Melting Pt: –112 °C Density: 1.104 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000719 CAS Number: 75-36-5 EINECS: 200-865-6 UN: 1717 ADR: 3,II Merck Index: 14,85 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...