Cobalt ≥99.995% (metals basis), slug, Puratronic®, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals
Danger

043226-QK 043226-QJ
AA43226-QKEA 529 USD
AA43226-QK AA43226-QJ
Cobalt ≥99.995% (metals basis), slug, Puratronic®, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Cobalt

Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈1.8g/slug Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR