Zirconyl nitrate hydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate
043224-14
043224-36
043224-22
AA43224-14EA
82.94
USD
AA43224-14
AA43224-36
AA43224-22
Zirconyl nitrate hydrate ≥99.9% (metals basis)
Zirconyl nitrate hydrate
Purity: 99.9%
CAS No.: 14985-18-3
Molecular Formula: ZrO(NO3)2.xH2O
Formula Weight: 231.23(anhy)
Physical Form: Crystalline
Odor: Odorless
UN No.: 1477
MDL No.: MFCD00149896
Formula:
N₂O₇Zr·H₂O Density: 2.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149896 CAS Number: 14985-18-3 EINECS: 237-529-3 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...