Graphite (natural) ≥99.8% (metals basis), soluble -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

043209-18 043209-30 043209-A1
AA43209-18EA 46.2 USD
AA43209-18 AA43209-30 AA43209-A1
Graphite (natural) ≥99.8% (metals basis), soluble -325 mesh
Graphite

Purity: 99.8% CAS No.: 7782-42-5 Physical Form: Flakes Appearance: Black Odor: Odorless UN No.: 1325 MDL No.: MFCD00144065

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1 III
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR