Benzyl acrylate 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acrylic Acid Benzyl Ester
Stabilization:
150 ppm Stabilised with 4-methoxyphenol (MEHQ)
043204-18
043204-30
043204-09
AA43204-18EA
154
USD
AA43204-18
AA43204-30
AAA43204-09
Benzyl acrylate 98% stabilised
Benzyl acrylate
Stab. with ca 150ppm 4-methoxyphenol
Preparation of high refractive index polyacrylates
Soluble in organic solvents
Applications: Preparation of high refractive index polyacrylates
Applications: Preparation of high refractive index polyacrylates
Formula:
C₁₀H₁₀O₂ MW: 162.19 g/mol Boiling Pt: 111 °C (8 mmHg) Density: 1.06 Storage Temperature: Ambient |
MDL Number:
MFCD00048147 CAS Number: 2495-35-4 EINECS: 219-673-9 UN: 3082 ADR: 9 III |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...