Carbon ≥99%, felt, Thickness 2.54 cm (1.0 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Charcoal

043201-RF
AA43201-RFEA 152 USD
AA43201-RF AA43201-RR
Carbon ≥99%, felt, Thickness 2.54 cm (1.0 in)
Carbon

~19.1g/10x10cm


Purity: 99.0% Physical Form: ≈19.1g/10x10cm MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Melting Pt: 3727
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-955-3
Merck Index: 13,01818

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR