Carbon ≥99%, felt, Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Charcoal

043199-RY 043199-RR
AA43199-RYEA 83.92 USD
AA43199-RY AA43199-RR
Carbon ≥99%, felt, Thickness 3.18 mm (0.125 in)
Carbon

MDL: MFCD00133992 Notes: Processed at 1400°C


Purity: 99.0% Physical Form: ≈2.4g/10x10cm MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Boiling Pt: ∼4000 °C (1013 hPa)
Melting Pt: 3727
Density: 3.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR