Acetonitrile, anhydrous ≥99.8%, ChemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
Packaged under argon in resealable ChemSeal bottle
Purity: 99.8+%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odor: Pungent
Melting Point: -48 to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878
Formula:
H₃CCN Boiling Pt: 80…82 °C Melting Pt: –48…–45 °C Density: 0.786 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 14,70 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...