Scandium, Oil Based Standard Solution, Specpure®, Sc 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
043129-18
AA43129-18EA
124.08
USD
AA43129-18
Scandium, Oil Based Standard Solution, Specpure®, Sc 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Scandium Standards
Physical Form: Liquid
MDL No.: MFCD00016323
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...