Chromium ≥99.99% (metals basis), pieces 1-25 mm (0.04-1 in)
Supplier: Thermo Scientific Chemicals
043066-18
043066-A1
043066-30
AA43066-18EA
119
USD
AA43066-18
AA43066-A1
AA43066-30
Chromium ≥99.99% (metals basis), pieces 1-25 mm (0.04-1 in)
Chromium
MDL: MFCD00010944
Purity: 99.99%
CAS No.: 7440-47-3
Physical Form: Broken plate
Odor: Odorless
MDL No.: MFCD00010944
Formula:
Cr Boiling Pt: 2469 °C (1013 hPa) Melting Pt: 1840 °C Density: 7.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 Merck Index: 12,02288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...