Calcium sulfate, granular, Drierite™ 6 mm (4 mesh) for drying, with indicator (cobalt (II) chloride)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite

043065-P2 043065-P5
AA43065-P2EA 78.67 USD
AA43065-P2 AA43065-P5
Calcium sulfate, granular, Drierite™ 6 mm (4 mesh) for drying, with indicator (cobalt (II) chloride)
Calcium sulphate

CAS No.: 7778-18-9 Physical Form: Granular, -4+6 Mesh Sensitivity: Hygroscopic Appearance: Blue to pink Odor: Odorless Merck Reference: 14,1706 MDL No.: MFCD00010912

Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
UN: 3077
ADR: 9 III
Merck Index: 14,1706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR