Germanium(IV) ethoxide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate

043052-14 043052-03 043052-06
AA43052-14EA 783 USD
AA43052-14 AA43052-03 AA43052-06
Germanium(IV) ethoxide ≥99.995% (metals basis)
Germanium (IV) ethoxide

Preparation of GeO2 by CVD; intermediate in sol-gel synthesis


Purity: 99.995% CAS No.: 14165-55-0 Molecular Formula: C8H20GeO4 Formula Weight: 252.84 Flash Point: 49°(121°F) Physical Form: Liquid Density: 1.14 Refractive Index: 1.4070 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Alcohol-like Melting Point: -72° Boiling Point: 185.5° UN No.: 1993 MDL No.: MFCD00015121


Preparation of GeO2 by CVD; intermediate in sol-gel synthesis

Formula: C₈H₂₀GeO₄
Boiling Pt: 185.5 °C
Melting Pt: –72 °C
Density: 1.14
Flash Pt: 49 °C (121 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015121
CAS Number: 14165-55-0
EINECS: 238-007-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR