Boron carbide 99+%

Supplier: Thermo Scientific Chemicals
Warning

043002-36
AA43002-36EA 194 USD
AA43002-36
Boron carbide 99+%
Boron carbide

MDL: MFCD00011520


Purity: 99+% CAS No.: 12069-32-8 Molecular Formula: B4C Formula Weight: 55.26 Physical Form: 10 Micron Powder Density: 2.52 Melting Point: 2350° Merck Reference: 14,1344 MDL No.: MFCD00011520

Formula: CB₄
MW: 55.25
Boiling Pt: ∼3500 °C (1013 hPa)
Melting Pt: 2350
Density: 2.52
Storage Temperature: Ambient
MDL Number: MFCD00011520
CAS Number: 12069-32-8
EINECS: 235-111-5
Merck Index: 14,1344

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR