Copper(II) sulfide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Copper sulfide, Cupric sulfide, Copper sulphide, Cupric sulphide, Copper(II)sulfide, Copper(II)sulphide

042925-30 042925-A1 042925-18
AA42925-30EA 248 USD
AA42925-30 AA42925-A1 AA42925-18
Copper(II) sulfide ≥99.8% (metals basis)
Copper (II) sulphide

MDL: MFCD00016066


Purity: 99.8% CAS No.: 1317-40-4 Molecular Formula: CuS Formula Weight: 95.60 Physical Form: -200 Mesh Powder Density: 3.9-4.6 Refractive Index: 1.45 Sensitivity: Air and Moisture Sensitive Melting Point: 220° dec. Merck Reference: 14,2655 MDL No.: MFCD00016066

Formula: CuS
Melting Pt: 220 °C (dec.)
Density: 3.9…4.6
Storage Temperature: Ambient
MDL Number: MFCD00016066
CAS Number: 1317-40-4
EINECS: 215-271-2
Merck Index: 14,2655

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR