Glassy carbon, rods, Diameter 3 mm (0.1 in) Type 1

Supplier: Thermo Scientific Chemicals

042822-DJ 042822-DM 042822-DR
AA42822-DJEA 191 USD
AA42822-DJ AA42822-DM AA42822-DR
Glassy carbon, rods, Diameter 3 mm (0.1 in) Type 1
Glassy carbon

MDL: MFCD00133992


Physical Form: ≈0.11g/cm MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR