Hexachloroplatinic(IV) acid hexahydrate Pt ≥ 37,5% ≥99.95% (metals basis), Premion® low nitrate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate
Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.
Light sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.
Formula:
H₂PtCl₆·6H₂O Melting Pt: 60 °C (dec.) Density: 2.430 Storage Temperature: Ambient |
MDL Number:
MFCD00149910 CAS Number: 18497-13-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III Merck Index: 14,7526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...