Hexachloroplatinic(IV) acid hexahydrate Pt ≥ 37,5% ≥99.95% (metals basis), Premion® low nitrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate

042815-03 042815-06 042815-14
AA42815-03EA 132.55 USD
AA42815-03 AA42815-06 AA42815-14
Hexachloroplatinic(IV) acid hexahydrate Pt ≥ 37,5% ≥99.95% (metals basis), Premion® low nitrate
Hexachloroplatinic (IV) acid hexahydrate

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.


Light sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.

Formula: H₂PtCl₆·6H₂O
Melting Pt: 60 °C (dec.)
Density: 2.430
Storage Temperature: Ambient
MDL Number: MFCD00149910
CAS Number: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 14,7526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR