Zinc chloride 1 M in diethyl ether
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
Packaged under argon in resealable ChemSeal™ bottles.
042781-AE
AA42781-AEEA
75.21
USD
AA42781-AE
AA42781-AP
Zinc chloride 1 M in diethyl ether
Zinc chloride
CAS No.: 7646-85-7
Molecular Formula: Cl2Zn
Formula Weight: 136.29
Flash Point: -40°(-40°F)
Physical Form: Liquid
Density: 0.835
Appearance: Colorless
Merck Reference: 14,10132
UN No.: 2924
MDL No.: MFCD00011295
Formula:
Cl₂Zn Density: 0.835 Flash Pt: –40 °C (–40 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 UN: 2924 ADR: 3,I Merck Index: 14,10132 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...