Silica gel 60 0.060-0.2mm (70-230 mesh), with fluorescent indicator

Supplier: Thermo Scientific Chemicals
Warning

042757-30 042757-A1 042757-A7 042757-18
AA42757-30EA 76 USD
AA42757-30 AA42757-A1 AA42757-A7 AA42757-18
Silica gel 60 0.060-0.2mm (70-230 mesh), with fluorescent indicator
Silica gel

pH 6.5-7.5


Applications: Dry column chromatography without applied pressure (gravity pulled)


Applications: Dry column chromatography without applied pressure (gravity pulled)

Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR