Silica gel, Pore size: 150 Å, wide pore
Supplier: Thermo Scientific Chemicals
|
Warning
|
Column chromatography without applied pressure (gravity pulled)
CAS No.: 63231-67-4
Physical Form: -100+200 Mesh, S.A. 350-400m2/g
Odor: Odorless
MDL No.: MFCD00011232
Column chromatography without applied pressure (gravity pulled)
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...