Silica gel, Pore size: 60 Å 0.075-0.2 mm (70-200 mesh)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Classic column chromatography with no applied pressure (gravity pulled)
CAS No.: 7631-86-9
Physical Form: -70+200 Mesh Powder, S.A. 470-570m2/g
Density: 2.4
Appearance: White
Odor: Odorless
pH: 6
Melting Point: 1710°
Boiling Point: 2230°
MDL No.: MFCD00011232
Classic column chromatography with no applied pressure (gravity pulled)
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.4 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...