Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
042687-FF
042687-FI
AA42687-FFEA
189
USD
AA42687-FF
AA42687-FI
Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.025 mm (0.001 in)
Holmium
MDL: MFCD00011049
Purity: 99.9%
CAS No.: 7440-60-0
Physical Form: ≈0.14g/25x25mm, Packaged in mylar under argon
Appearance: Gray to silver
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00011049
Formula:
Ho Storage Temperature: Ambient |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 UN: 3178 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...