Ammonium dihydrogen phosphate 10% in nitric acid 2%, Specpure® matrix modifier solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

042663-AE
AA42663-AEEA 150 USD
AA42663-AE
Ammonium dihydrogen phosphate 10% in nitric acid 2%, Specpure® matrix modifier solution
Ammonium dihydrogen phosphate
Formula: O₃SrTi
MW: 115.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR