Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length 6.35 mm (0.25 in), Diameter 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

042558-14 042558-06
AA42558-14EA 274 USD
AA42558-14 AA42558-06
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length 6.35 mm (0.25 in), Diameter 3.175 mm (0.125 in)
Zirconium

MDL: MFCD00011303


Purity: 99.95% CAS No.: 7440-67-7 Physical Form: ≈0.33g/slug Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR