Calcium hypochlorite, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
042548-30
AA42548-18EA
41.84
USD
AA42548-18
AA42548-30
Calcium hypochlorite, tech.
Calcium hypochlorite
Deodorant, oxidizing agent, bleaching agent
CAS No.: 7778-54-3
Molecular Formula: CaCl2O2
Formula Weight: 142.98
Physical Form: Granular
Density: 2.350
Appearance: White to pale brown
Odor: Like chlorine
pH: 11,5
Melting Point: 100°
Merck Reference: 14,1674
UN No.: 1748
MDL No.: MFCD00010900
Deodorant, oxidizing agent, bleaching agent
Formula:
CaCl₂O₂ MW: 142.98 g/mol Melting Pt: 100 °C Density: 2.350 Storage Temperature: Ambient |
MDL Number:
MFCD00010900 CAS Number: 7778-54-3 EINECS: 231-908-7 UN: 1748 ADR: 5.1,II Merck Index: 14,1674 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...