Digallium trioxide ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

042528-14 042528-06 042528-22
AA42528-14EA 265 USD
AA42528-14 AA42528-06 AA42528-22
Digallium trioxide ≥99.999% (metals basis), Puratronic®
Digallium trioxide

MDL: MFCD00011020


Purity: 99.999% CAS No.: 12024-21-4 Molecular Formula: Ga2O3 Formula Weight: 187.44 Physical Form: -100 Mesh Powder Density: 6.44 Refractive Index: 1.92 Odor: Odorless Melting Point: 1900° Merck Reference: 14,4346 MDL No.: MFCD00011020

Formula: Ga₂O₃
MW: 187.44
Melting Pt: 1794.85
Density: 6.44
Storage Temperature: Ambient
MDL Number: MFCD00011020
CAS Number: 12024-21-4
EINECS: 234-691-7
Merck Index: 14,4346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR