n-Octacosane 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-octacosane
042460-30
042460-09
042460-18
AA42460-30EA
342
USD
AA42460-30
AA42460-09
AA42460-18
n-Octacosane 99%
n-Octacosane
MDL: MFCD00009355
Purity: 99%
CAS No.: 630-02-4
Molecular Formula: CH3(CH2)26CH3
Formula Weight: 394.77
Flash Point: 227°(441°F)
Physical Form: Crystalline
Melting Point: 61-63°
Boiling Point: 440°
MDL No.: MFCD00009355
Formula:
H₃C(CH₂)₂₆CH₃ MW: 394.77 g/mol Boiling Pt: 432 Melting Pt: 61…63 °C Density: 0.8067 g/cm³ (20 °C) Flash Pt: 227 °C (441 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009355 CAS Number: 630-02-4 EINECS: 211-125-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...