Vanadium, Oil Based Standard Solution, Specpure®, V 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042452-18
AA42452-18EA
85.4
USD
AA42452-18
Vanadium, Oil Based Standard Solution, Specpure®, V 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Vanadium Standards
Physical Form: Liquid
MDL No.: MFCD00011453
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...