Titanium, Oil Based Standard Solution, Specpure®, Ti 5000µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042451-18
AA42451-18EA
74.8
USD
AA42451-18
Titanium, Oil Based Standard Solution, Specpure®, Ti 5000µg/g, Thermo Scientific
Standards
Cationic Standards
Titanium Standards
Physical Form: Liquid
MDL No.: MFCD00011264
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...