Tin, Oil based Standard Solution, Specpure®, Sn 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042450-18
AA42450-18EA
74.2
USD
AA42450-18
Tin, Oil based Standard Solution, Specpure®, Sn 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Tin Standards
Physical Form: Liquid
MDL No.: MFCD00133862
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...