Potassium, Oil based standard solution, Specpure®, K 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042448-18
AA42448-18EA
76.8
USD
AA42448-18
Potassium, Oil based standard solution, Specpure®, K 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Potassium Standards
Physical Form: Liquid
MDL No.: MFCD00133776
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...