Nickel, Oil Based Standard Solution, Specpure®, Ni 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042446-18
AA42446-18EA
87.9
USD
AA42446-18
Nickel, Oil Based Standard Solution, Specpure®, Ni 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Nickel Standards
Physical Form: Liquid
MDL No.: MFCD00011137
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...