Molybdenum, Oil Based Standard Solution, Specpure®, Mo 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042445-18
AA42445-18EA
75.31
USD
AA42445-18
Molybdenum, Oil Based Standard Solution, Specpure®, Mo 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Molybdenum Standards
Physical Form: Liquid
MDL No.: MFCD00003465
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...