Barium, Oil based standard solution, Specpure®, Ba 5000µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042434-18
AA42434-18EA
71.25
USD
AA42434-18
Barium, Oil based standard solution, Specpure®, Ba 5000µg/g, Thermo Scientific
Standards
Cationic Standards
Barium Standards
Physical Form: Liquid
MDL No.: MFCD00134031
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...